Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxypivalic acid 97%

Supplier: AMBEED, INC
Danger

A128522-100G A128522-10G A128522-25G
77109-052EA 39.15 USD InStock
77109-052 77109-054 77298-310
Hydroxypivalic acid 97%
Hydroxypivalic acid
Precaución: for Research Use Only


Formula: C₅H₁₀O₃
Boiling Pt: 242.8 °C (760 mm Hg)
Melting Pt: 123…127 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00059953
CAS Number: 4835-90-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty