Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-4-(trifluoromethyl)benzoic acid 95%

Supplier: AMBEED, INC
Warning

A218739-1G A218739-5G A218739-100MG A218739-10G A218739-250MG A218739-1G
103818-740EA 158 USD InStock
103818-740 103849-610 76986-990 76986-992 76986-994 77088-138
3-Hydroxy-4-(trifluoromethyl)benzoic acid 95%
3-Hydroxy-4-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 305.5±42.0 °C (760 mm Hg)
Flash Pt: 138.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16999826
CAS Number: 126541-87-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty