Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxycyclopentanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A366398-1G A366398-250MG A366398-1G
103826-398EA 76 USD InStock
103826-398 76817-442 77306-772
3-Hydroxycyclopentanecarboxylic acid 95%
3-Hydroxycyclopentanecarboxylic acid
Precaución: for Research Use Only


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 297.2±33.0 °C (760 mm Hg)
Flash Pt: 147.8 °C
Storage Temperature: Ambient
MDL Number: MFCD13659410
CAS Number: 101080-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty