Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxypyridine-4-carboxaldehyde 97%

Supplier: AMBEED, INC
Warning

A201966-1G A201966-5G A201966-10G A201966-250MG A201966-5G
103817-344EA 44 USD InStock
103817-344 103847-980 76999-366 76999-368 77302-784
3-Hydroxypyridine-4-carboxaldehyde 97%
3-Hydroxypyridine-4-carboxaldehyde
Precaución: for Research Use Only


Formula: C6H5NO2
MW: 123.1094 g/mol
Boiling Pt: 323.8 °C (760 mm Hg)
Flash Pt: 149.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318565
CAS Number: 1849-54-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty