Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodo-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A362440-1G A362440-25G A362440-5G A362440-10G A362440-250MG
103826-318EA 7 USD InStock
103826-318 103837-334 76840-548 76840-550 76840-552 77003-374 77003-376
3-Iodo-1H-pyrazole 98%
3-Iodo-1H-pyrazole
Precaución: for Research Use Only


Formula: C₃H₃IN₂
MW: 193.98 g/mol
Boiling Pt: 291.9 °C (760 mm Hg)
Melting Pt: 74…75 °C
Flash Pt: 130.3 °C
Storage Temperature: Ambient
MDL Number: MFCD12022324
CAS Number: 4522-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty