Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Iodobenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A135649-1G A135649-10G A135649-250MG A135649-5G A135649-1G A135649-25G
103802-190EA 6 USD InStock
103802-190 77137-062 77137-064 77298-804 77315-032 77315-034
3-Iodobenzaldehyde 97%
3-Iodobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅IO
MW: 232.02
Boiling Pt: 266.1 °C (760 mm Hg)
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00039573
CAS Number: 696-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty