Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine 98%

Supplier: AMBEED, INC
Warning

A117729-1G A117729-100MG A117729-250MG A117729-1G
103798-932EA 34 USD InStock
103798-932 103824-822 77271-310 77271-312 77311-600
[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine 98%
[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine
Precaución: for Research Use Only


Formula: C₆H₁₀N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06335179
CAS Number: 88786-11-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty