Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-2-methyl-3-oxopropanoic acid 95%

Supplier: AMBEED, INC
Warning

A106365-1G A106365-100MG A106365-250MG
103797-028EA 113 USD InStock
103797-028 77160-482 77160-484
3-Methoxy-2-methyl-3-oxopropanoic acid 95%
3-Methoxy-2-methyl-3-oxopropanoic acid
Precaución: for Research Use Only


Formula: C₅H₈O₄
MW: 132.11 g/mol
Boiling Pt: 227.3 °C (760 mm Hg)
Flash Pt: 95.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00154582
CAS Number: 3097-74-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty