Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Methoxy-4-methylphenyl)acetonitrile 97%

Supplier: AMBEED, INC
Warning

A246373-1G A246373-5G A246373-1G A246373-25G A246373-10G A246373-5G
103811-706EA 13 USD InStock
103811-706 103851-538 77090-834 77090-836 77276-196 77304-388
(3-Methoxy-4-methylphenyl)acetonitrile 97%
(3-Methoxy-4-methylphenyl)acetonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO
MW: 161.2004 g/mol
Boiling Pt: 277.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00060309
CAS Number: 64829-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty