Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-4-nitrobenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A219338-1G A219338-5G A219338-1G A219338-5G A219338-250MG
103818-782EA 44 USD InStock
103818-782 103849-658 77088-210 77088-212 77275-440
3-Methoxy-4-nitrobenzaldehyde 95%
3-Methoxy-4-nitrobenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇NO₄
MW: 181.1455 g/mol
Boiling Pt: 334.8 °C (760 mm Hg)
Flash Pt: 171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08703367
CAS Number: 80410-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty