Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethyl)-m-anisidine 97%

Supplier: AMBEED, INC
Warning

A312490-1G A312490-5G A312490-1G A312490-25G A312490-5G A312490-10G
103814-698EA 9 USD InStock
103814-698 103835-888 76838-694 76838-696 76838-698 77251-008
5-(Trifluoromethyl)-m-anisidine 97%
5-(Trifluoromethyl)-m-anisidine
Precaución: for Research Use Only


Formula: C₈H₈F₃NO
Boiling Pt: 242 °C (760 mm Hg)
Melting Pt: 50…52 °C
Flash Pt: 100.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000393
CAS Number: 349-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty