Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-5-methylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A253581-1G A253581-5G A253581-250MG A253581-1G A253581-5G
103812-098EA 17 USD InStock
103812-098 103851-948 76815-738 77091-384 77091-386
3-Methoxy-5-methylbenzaldehyde 95%
3-Methoxy-5-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11046639
CAS Number: 90674-26-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty