Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-6-(trifluoromethyl)picolinic acid 98%

Supplier: AMBEED, INC
Warning

A689990-1G A689990-250MG
103840-574EA 1635 USD InStock
103840-574 76844-792 76992-648
3-Methoxy-6-(trifluoromethyl)picolinic acid 98%
3-Methoxy-6-(trifluoromethyl)picolinic acid
Precaución: for Research Use Only


Formula: C₈H₆F₃NO₃
MW: 221.13 g/mol
Boiling Pt: 299.4±40.0 °C (760 mm Hg)
Flash Pt: 134.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15478069
CAS Number: 1214330-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty