Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Methoxyphenyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A382004-5G A382004-100G A382004-10G A382004-500G A382004-25G
103837-744EA 6 USD InStock
103837-744 76841-064 77251-900 77251-902 77251-904 77306-928
(3-Methoxyphenyl)acetic acid 97%
(3-Methoxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄(OCH₃))CH₂COOH
Boiling Pt: 305.5 °C (760 mm Hg)
Melting Pt: 65…69 °C
Flash Pt: 124.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004334
CAS Number: 1798-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty