Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxypyridine-2-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A118807-1G A118807-250MG A118807-100MG
103799-110EA 97 USD InStock
103799-110 77670-672 77880-962
3-Methoxypyridine-2-carbaldehyde 97%
3-Methoxypyridine-2-carbaldehyde
Precaución: Inert atmosphere, store in freezer, under –20 °C.


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 239.4 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD05664031
CAS Number: 1849-53-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty