Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1-phenyl-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A169819-1G A169819-5G A169819-10G A169819-250MG A169819-1G
103806-416EA 21 USD InStock
103806-416 103844-680 76690-130 76690-132 77301-002
3-Methyl-1-phenyl-1H-pyrazole 95%
3-Methyl-1-phenyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 255 °C (760 mm Hg)
Melting Pt: 34…36 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003123
CAS Number: 1128-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty