Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-2-phenylpyridine 95%

Supplier: AMBEED, INC
Warning

A247484-5G A247484-10G A247484-250MG A247484-25G A247484-1G A247484-5G
103811-772EA 21 USD InStock
103811-772 103851-608 77031-406 77031-408 77031-410 77090-946 77304-414
3-Methyl-2-phenylpyridine 95%
3-Methyl-2-phenylpyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 148
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00006283
CAS Number: 10273-90-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty