Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%

Supplier: AMBEED, INC
Warning

A231085-1G A231085-5G A231085-10G A231085-250MG A231085-1G A231085-5G
103810-752EA 21 USD InStock
103810-752 103850-498 76815-140 76815-142 77089-370 77089-372
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₉BO₄
Boiling Pt: 394.1±35.0 °C (760 mm Hg)
Flash Pt: 192.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08061640
CAS Number: 269409-74-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty