Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-5-nitrobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A213161-1G A213161-5G A213161-10G A213161-1G A213161-5G
103818-266EA 13 USD InStock
103818-266 103848-970 76814-646 77087-454 77303-272
3-Methyl-5-nitrobenzoic acid 95%
3-Methyl-5-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₄
Melting Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00129754
CAS Number: 113882-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty