Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-3-methyl-5-nitropyridine 98%

Supplier: AMBEED, INC
Warning

A431757-1G A431757-5G A431757-1G A431757-250MG
103827-528EA 26 USD InStock
103827-528 103838-594 76842-286 76877-278
2-Cyano-3-methyl-5-nitropyridine 98%
2-Cyano-3-methyl-5-nitropyridine
Precaución: for Research Use Only


Formula: C₇H₅N₃O₂
Boiling Pt: 366.6 °C (760 mm Hg)
Flash Pt: 175.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07375120
CAS Number: 65169-63-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty