Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylsalicylaldehyde 98%

Supplier: AMBEED, INC
Warning

A153888-1G A153888-5G A153888-100G A153888-1G A153888-25G A153888-5G A153888-10G
103804-578EA 16 USD InStock
103804-578 103842-756 77247-386 77318-440 77318-442 77318-444 77746-059
3-Methylsalicylaldehyde 98%
3-Methylsalicylaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₂
Boiling Pt: 204 °C (760 mm Hg)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00040798
CAS Number: 824-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty