Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Morpholino-1-(4-nitrophenyl)-5,6-dihydropyridin-2(1H)-one 95%

Supplier: AMBEED, INC
Warning

A206207-1G A206207-5G A206207-10G A206207-1G A206207-25G A206207-5G
103817-692EA 32 USD InStock
103817-692 103848-362 76825-744 77086-604 77086-606 77086-608
3-Morpholino-1-(4-nitrophenyl)-5,6-dihydropyridin-2(1H)-one 95%
3-Morpholino-1-(4-nitrophenyl)-5,6-dihydropyridin-2(1H)-one
Precaución: for Research Use Only


Formula: C₁₅H₁₇N₃O₄
Boiling Pt: 506.532 °C (760 mm Hg)
Flash Pt: 260.141 °C
Storage Temperature: Freezer
MDL Number: MFCD19440872
CAS Number: 503615-03-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty