Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Morpholin-4-ylpropionic acid 98%

Supplier: AMBEED, INC
Warning

A111981-1G A111981-5G A111981-1G A111981-25G
103797-932EA 12 USD InStock
103797-932 103823-756 77284-196 77284-198
3-Morpholin-4-ylpropionic acid 98%
3-Morpholin-4-ylpropionic acid
Precaución: for Research Use Only


Formula: C₇H₁₃NO₃
Boiling Pt: 299.2 °C (760 mm Hg)
Melting Pt: 217 °C
Flash Pt: 134.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02089311
CAS Number: 4497-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty