Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitrobenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A192817-1000G A192817-100G A192817-25G A192817-500G
103809-280EA 46 USD InStock
103809-280 77084-972 77111-838 77111-840 77111-842
3-Nitrobenzaldehyde 97%
3-Nitrobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 232
Melting Pt: 58
Storage Temperature: Ambient
MDL Number: MFCD00007249
CAS Number: 99-61-6
Merck Index: 12,06684
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty