Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitroisonicotinic acid 95%

Supplier: AMBEED, INC
Warning

A402966-1G A402966-5G A402966-100MG A402966-250MG
103827-084EA 31 USD InStock
103827-084 103838-124 76841-594 76841-596 77291-160 77291-162
3-Nitroisonicotinic acid 95%
3-Nitroisonicotinic acid
Precaución: for Research Use Only


Formula: C6H4N2O4
MW: 168.11 g/mol
Boiling Pt: 467.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04114244
CAS Number: 59290-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty