Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A127229-100G A127229-10G A127229-25G A127229-5G
77271-836EA 79 USD InStock
77271-836 77271-838 77313-180 77313-182
2-Hydroxy-3-nitropyridine 97%
2-Hydroxy-3-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₄N₂O₃
MW: 140.1
Boiling Pt: 368.8 °C (760 mm Hg)
Melting Pt: 224
Flash Pt: 176.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006270
CAS Number: 6332-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty