Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxocyclohexanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A327379-1G A327379-5G A327379-100MG A327379-250MG
103825-464EA 47 USD InStock
103825-464 103836-414 77290-370 77290-372
3-Oxocyclohexanecarboxylic acid 95%
3-Oxocyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₁₀O₃
MW: 142.1525 g/mol
Boiling Pt: 309.7 °C (760 mm Hg)
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD01320172
CAS Number: 16205-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty