Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxoisoindoline-5-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A235780-1G A235780-5G A235780-100MG A235780-250MG
103811-032EA 208 USD InStock
103811-032 103850-800 77881-614 77881-615
3-Oxoisoindoline-5-carbonitrile 95%
3-Oxoisoindoline-5-carbonitrile
Precaución: Sealed in dry, at room temperature.


Formula: C₉H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD12924851
CAS Number: 1261726-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty