Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenoxypropionic acid 98%

Supplier: AMBEED, INC
Warning

A154460-5G A154460-100G A154460-1G A154460-25G A154460-5G
103842-830EA 9 USD InStock
103842-830 77273-090 77273-092 77318-554 77318-556
3-Phenoxypropionic acid 98%
3-Phenoxypropionic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.18
Boiling Pt: 235
Melting Pt: 92
Flash Pt: 107.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002769
CAS Number: 7170-38-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty