Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenylmaleic anhydride 97%

Supplier: AMBEED, INC
Warning

A178809-1G A178809-5G A178809-10G A178809-250MG A178809-1G A178809-25G
103807-368EA 9 USD InStock
103807-368 103845-708 76799-638 76799-640 77083-148 77083-150
Phenylmaleic anhydride 97%
Phenylmaleic anhydride
Precaución: for Research Use Only


Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Boiling Pt: 341.6 °C (760 mm Hg)
Melting Pt: 120…122 °C
Flash Pt: 167.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00015472
CAS Number: 36122-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty