Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenylpyrazin-2-ol 97%

Supplier: AMBEED, INC
Warning

A210557-1G A210557-5G A210557-100MG A210557-250MG A210557-1G A210557-5G
103799-488EA 30 USD InStock
103799-488 103818-018 103848-712 76873-658 76873-660 77087-090 77087-092
3-Phenylpyrazin-2-ol 97%
3-Phenylpyrazin-2-ol
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O
Boiling Pt: 457.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD12828231
CAS Number: 73200-73-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty