Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyrazolidinone hydrochloride 97%

Supplier: AMBEED, INC
Warning

A152000-1G A152000-5G A152000-250MG A152000-1G A152000-5G
103804-336EA 14 USD InStock
103804-336 103842-486 77286-140 77318-084 77318-086
3-Pyrazolidinone hydrochloride 97%
3-Pyrazolidinone hydrochloride
Precaución: for Research Use Only


Formula: C₃H₇ClN₂O
Melting Pt: 211…212 °C
Storage Temperature: Ambient
MDL Number: MFCD08594075
CAS Number: 1752-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty