Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(tert-Butoxycarbonyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A125543-1G A125543-100G A125543-10G A125543-25G A125543-5G A125543-1G
103800-692EA 5 USD InStock
103800-692 76797-652 76797-654 76797-656 77298-058 77312-840
3-(tert-Butoxycarbonyl)phenylboronic acid 98%
3-(tert-Butoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅BO₄
MW: 222.05 g/mol
Boiling Pt: 375 °C (760 mm Hg)
Melting Pt: 96…102 °C
Flash Pt: 180.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01630855
CAS Number: 220210-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty