Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thiopheneboronic acid 98%

Supplier: AMBEED, INC
Warning

A209390-5G A209390-25G A209390-100G A209390-500G
103817-936EA 6 USD InStock
103817-936 103848-622 77086-964 77287-918 77287-920
3-Thiopheneboronic acid 98%
3-Thiopheneboronic acid
Precaución: for Research Use Only


Formula: C₄H₅BO₂S
MW: 127.96
Boiling Pt: 287.9 °C (760 mm Hg)
Melting Pt: 164…169 °C
Flash Pt: 127.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00151851
CAS Number: 6165-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty