Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Nitroanilino)phenol 98% (by GC)

Supplier: AMBEED, INC
Warning

A112970-1G A112970-100G A112970-10G A112970-250MG A112970-25G A112970-500G A112970-1G A112970-5G
103798-142EA 23 USD InStock
103798-142 103823-976 76765-986 76765-988 76765-990 76765-992 76765-994 77297-218 77297-220
4-(2-Nitroanilino)phenol 98% (by GC)
4-(2-Nitroanilino)phenol
Precaución: for Research Use Only


Formula: C₁₂H₁₀N₂O₃
MW: 230.22 g/mol
Boiling Pt: 384.9 °C (760 mm Hg)
Melting Pt: 143…145 °C
Flash Pt: 186.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00239470
CAS Number: 54381-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty