Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((4-Ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)aniline 97%

Supplier: AMBEED, INC
Warning

A222661-1G A222661-5G A222661-10G A222661-250MG A222661-1G A222661-5G
103818-988EA 105 USD InStock
103818-988 103849-866 76826-224 76826-226 77088-478 77088-480
4-((4-Ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)aniline 97%
4-((4-Ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₁₄H₂₀F₃N₃
Storage Temperature: Refrigerator
MDL Number: MFCD09909923
CAS Number: 630125-91-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty