Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((6-Bromopyridin-3-yl)methyl)morpholine 95%

Supplier: AMBEED, INC
Warning

A100731-1G A100731-5G A100731-100MG A100731-250MG A100731-1G
103795-598EA 52 USD InStock
103795-598 103821-294 76982-150 76982-152 77296-830
4-((6-Bromopyridin-3-yl)methyl)morpholine 95%
4-((6-Bromopyridin-3-yl)methyl)morpholine
Precaución: for Research Use Only


Formula: C₁₀H₁₃BrN₂O
Boiling Pt: 342.438 °C (760 mm Hg)
Flash Pt: 160.901 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11846170
CAS Number: 364793-93-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty