Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A198410-1G A198410-5G A198410-100MG A198410-250MG A198410-1G A198410-5G A198410-10G
103817-012EA 36 USD InStock
103817-012 103847-638 76825-470 76825-472 77085-660 77085-662 77683-547
4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid 97%, Ambeed.Inc
4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid
Precaución: For research use only.


Formula: C₂₂H₁₅N₃O₂
MW: 353.38 g/mol
Boiling Pt: 560.8 °C (760 mm Hg)
Flash Pt: 292.9 °C
Storage Temperature: Ambient
MDL Number: MFCD22056799
CAS Number: 158014-74-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty