Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1-Boc-piperidine-4-yl)butanoic acid 97%

Supplier: AMBEED, INC
Warning

A137503-1G A137503-10G A137503-250MG A137503-1G A137503-25G A137503-5G
103802-464EA 18 USD InStock
103802-464 76782-744 76782-746 77315-430 77315-432 77315-434
4-(1-Boc-piperidine-4-yl)butanoic acid 97%
4-(1-Boc-piperidine-4-yl)butanoic acid
Precaución: for Research Use Only


Formula: C₁₄H₂₅NO₄
MW: 271.36 g/mol
Boiling Pt: 399.7 °C (760 mm Hg)
Melting Pt: 70…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114965
CAS Number: 142247-38-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty