Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1H-Pyrazol-1-yl)piperidine 98%

Supplier: AMBEED, INC
Warning

A224627-1G A224627-5G A224627-250MG A224627-1G A224627-5G
103819-126EA 165 USD InStock
103819-126 103850-018 76814-996 77303-704 77303-706
4-(1H-Pyrazol-1-yl)piperidine 98%
4-(1H-Pyrazol-1-yl)piperidine
Precaución: for Research Use Only


Formula: C₈H₁₃N₃
Storage Temperature: Ambient
MDL Number: MFCD09998987
CAS Number: 762240-09-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty