Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Bromoethyl)benzonitrile 97%

Supplier: AMBEED, INC
Warning

A126530-1G A126530-100MG A126530-250MG A126530-1G
103800-826EA 20 USD InStock
103800-826 76782-054 76782-056 77313-048
4-(2-Bromoethyl)benzonitrile 97%
4-(2-Bromoethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₉H₈BrN
Boiling Pt: 290 °C (760 mm Hg)
Melting Pt: 53…54 °C
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD12025077
CAS Number: 72054-56-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty