Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carboxycinnamic acid 95%

Supplier: AMBEED, INC
Warning

A103866-1G A103866-5G A103866-10G A103866-250MG A103866-25G A103866-1G A103866-5G
103796-618EA 15 USD InStock
103796-618 103822-328 76796-386 76796-388 76796-390 77309-194 77309-196
4-Carboxycinnamic acid 95%
4-Carboxycinnamic acid


Formula: C₁₀H₈O₄
Boiling Pt: 351.7±22.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00016543
CAS Number: 19675-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty