Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[2-(Morpholin-4-yl)ethyl]aniline 97%

Supplier: AMBEED, INC
Warning

A111728-1G A111728-10G A111728-250MG A111728-25G A111728-1G A111728-5G
103797-896EA 34 USD InStock
103797-896 103823-714 76982-756 76982-758 76982-760 77284-174 77284-176
4-[2-(Morpholin-4-yl)ethyl]aniline 97%
4-[2-(Morpholin-4-yl)ethyl]aniline
Precaución: for Research Use Only


Formula: C₁₂H₁₈N₂O
MW: 206.284 g/mol
Boiling Pt: 343.596 °C (760 mm Hg)
Flash Pt: 161.602 °C
Storage Temperature: Ambient
MDL Number: MFCD04114508
CAS Number: 262368-47-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty