Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3-Formylphenoxy)benzonitrile 97%

Supplier: AMBEED, INC
Warning

A100025-1G A100025-5G A100025-10G A100025-1G A100025-5G
103795-204EA 6 USD InStock
103795-204 103820-926 77245-042 77296-542 77296-544
4-(3-Formylphenoxy)benzonitrile 97%
4-(3-Formylphenoxy)benzonitrile
Precaución: for Research Use Only


Formula: C₁₄H₉NO₂
Boiling Pt: 408.7±25.0 °C (760 mm Hg)
Flash Pt: 178.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11500847
CAS Number: 90178-72-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty