Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-propanenitrile 95%

Supplier: AMBEED, INC
Warning

A155399-1G A155399-5G A155399-250MG A155399-1G A155399-5G
103804-756EA 130 USD InStock
103804-756 103842-952 76973-286 77318-666 77318-668
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-propanenitrile 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-propanenitrile
Precaución: for Research Use Only


Formula: C₁₂H₁₈BN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD16660233
CAS Number: 1022092-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty