Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: AMBEED, INC
Warning

A125592-1G A125592-10G A125592-250MG A125592-25G A125592-5G A125592-1G
103800-702EA 20 USD InStock
103800-702 77026-762 77026-764 77298-070 77298-072 77312-848
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine 98%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₁₃H₁₇BN₂O₂
Boiling Pt: 405.7 °C (760 mm Hg)
Melting Pt: 250…254 °C
Flash Pt: 199.2 °C
Storage Temperature: Freezer
MDL Number: MFCD11616360
CAS Number: 942919-26-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty