Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enol 95%

Supplier: AMBEED, INC
Warning

A148073-1G A148073-100MG A148073-250MG A148073-1G
103803-872EA 248 USD InStock
103803-872 77150-304 77150-306 77299-632
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enol 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enol
Precaución: for Research Use Only


Formula: C₁₂H₂₁BO₃
Boiling Pt: 276.6±50.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD12025483
CAS Number: 1310384-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty