Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oxindole-4-boronic acid pinacol ester ≥95%

Supplier: AMBEED, INC
Warning

A181793-1G A181793-100MG A181793-250MG A181793-5G
103807-652EA 154 USD InStock
103807-652 77671-566 77881-377 77881-378
Oxindole-4-boronic acid pinacol ester ≥95%
Oxindole-4-boronic acid pinacol ester
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₁₄H₁₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11504971
CAS Number: 1150271-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty