Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxyphenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A131370-1G A131370-100G A131370-10G A131370-500G A131370-1G A131370-25G A131370-5G
103801-572EA 5 USD InStock
103801-572 76718-962 76718-964 76718-966 77314-072 77314-074 77314-076
4-Hydroxyphenylboronic acid pinacol ester 98%
4-Hydroxyphenylboronic acid pinacol ester


Formula: C₁₂H₁₇BO₃
Boiling Pt: 333.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02093756
CAS Number: 269409-70-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty