Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Bromobenzyl)morpholine 95%

Supplier: AMBEED, INC
Warning

A352998-1G A352998-25G A352998-5G A352998-100G A352998-10G
103826-082EA 12 USD InStock
103826-082 103837-084 76840-190 76840-192 76840-194 77290-680 77290-682
4-(4-Bromobenzyl)morpholine 95%
4-(4-Bromobenzyl)morpholine
Precaución: for Research Use Only


Formula: C₁₁H₁₄BrNO
Boiling Pt: 313.2 °C (760 mm Hg)
Flash Pt: 143.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00979535
CAS Number: 132833-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty